Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAAATCAATGGCTGGATGCAGA[A/G]GGACAAGTGATAGGACGGGGAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613337 MIM: 612264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371855 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371855 - uncharacterized LOC105371855 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH10 - membrane associated ring-CH-type finger 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100875.2 | Intron | NP_001094345.1 | ||||
NM_001288779.1 | Intron | NP_001275708.1 | ||||
NM_001288780.1 | Intron | NP_001275709.1 | ||||
NM_152598.3 | Intron | NP_689811.2 | ||||
XM_005257095.2 | Intron | XP_005257152.2 | ||||
XM_005257096.2 | Intron | XP_005257153.2 | ||||
XM_005257097.3 | Intron | XP_005257154.2 | ||||
XM_005257098.2 | Intron | XP_005257155.2 | ||||
XM_005257100.2 | Intron | XP_005257157.2 | ||||
XM_005257102.2 | Intron | XP_005257159.2 | ||||
XM_005257103.1 | Intron | XP_005257160.1 | ||||
XM_005257106.4 | Intron | XP_005257163.2 | ||||
XM_011524435.1 | Intron | XP_011522737.1 | ||||
XM_011524436.1 | Intron | XP_011522738.1 | ||||
XM_011524437.1 | Intron | XP_011522739.1 |
MRC2 - mannose receptor C type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |