Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATCCTTGGTTCTTGGGGCATGA[C/G]AGAAGAGAAAAGGTAAATATGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL20 - F-box and leucine rich repeat protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184906.1 | 8606 | UTR 3 | NP_001171835.1 | |||
NM_032875.2 | 8606 | UTR 3 | NP_116264.2 | |||
XM_005257746.3 | 8606 | UTR 3 | XP_005257803.2 | |||
XM_005257747.3 | 8606 | UTR 3 | XP_005257804.2 |
LOC101929578 - transcription factor CP2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |