Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTGGATTAGTATTCGGTGACT[A/C]TAATGCCATCAGGGTTTAGCTCGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610008 MIM: 188830 MIM: 609224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR635 - microRNA 635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAR1A - protein kinase cAMP-dependent type I regulatory subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276289.1 | Intron | NP_001263218.1 | ||||
NM_001276290.1 | Intron | NP_001263219.1 | ||||
NM_001278433.1 | Intron | NP_001265362.1 | ||||
NM_002734.4 | Intron | NP_002725.1 | ||||
NM_212471.2 | Intron | NP_997636.1 | ||||
NM_212472.2 | Intron | NP_997637.1 | ||||
XM_011524983.2 | Intron | XP_011523285.1 | ||||
XM_011524984.2 | Intron | XP_011523286.1 | ||||
XM_011524985.2 | Intron | XP_011523287.1 |
WIPI1 - WD repeat domain, phosphoinositide interacting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320772.1 | Intron | NP_001307701.1 | ||||
NM_017983.6 | Intron | NP_060453.3 | ||||
XM_017024808.1 | Intron | XP_016880297.1 |