Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGCTCTCTTTCCAGTGCGGT[A/G]TCCTTGGCCCTGGTGGGTGGAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRC5C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPRC5C - G protein-coupled receptor class C group 5 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018653.3 | Intron | NP_061123.3 | ||||
NM_022036.2 | Intron | NP_071319.2 | ||||
XM_005257512.1 | Intron | XP_005257569.1 | ||||
XM_005257514.1 | Intron | XP_005257571.1 | ||||
XM_017024855.1 | Intron | XP_016880344.1 | ||||
XM_017024856.1 | Intron | XP_016880345.1 |