Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCCATTCTTCTCACCAATAGC[C/T]ATGATGGACTGGATGGTGTAAGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602268 MIM: 605129 | ||||||||||||||||||||
Literature Links: |
AOC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AOC2 - amine oxidase, copper containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME3 - proteasome activator subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267045.1 | 716 | Missense Mutation | CAT,TAT | H,Y 87 | NP_001253974.1 | |
NM_005789.3 | 716 | Missense Mutation | CAT,TAT | H,Y 76 | NP_005780.2 | |
NM_176863.2 | 716 | Missense Mutation | CAT,TAT | H,Y 76 | NP_789839.1 | |
XM_011524177.2 | 716 | Missense Mutation | CAT,TAT | H,Y 15 | XP_011522479.1 |