Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGGTGTCCAGCTCCTGGCACTG[T/G]TAGAGTGCTACACTGTCCAACACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 113705 | ||||||||||||||||||||
Literature Links: |
BRCA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRCA1 - BRCA1, DNA repair associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007294.3 | 5830 | Nonsense Mutation | NP_009225.1 | |||
NM_007297.3 | 5830 | Nonsense Mutation | NP_009228.2 | |||
NM_007298.3 | 5830 | Nonsense Mutation | NP_009229.2 | |||
NM_007299.3 | 5830 | UTR 3 | NP_009230.2 | |||
NM_007300.3 | 5830 | Nonsense Mutation | NP_009231.2 |