Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAACAGGCCAAGATGAAACAC[C/G]AGGGGCACTCTTCACAGAAATTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJC1 - gap junction protein gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080383.1 | 3907 | UTR 3 | NP_001073852.1 | |||
NM_005497.3 | 3907 | UTR 3 | NP_005488.2 | |||
XM_005256920.1 | 3907 | UTR 3 | XP_005256977.1 | |||
XM_005256921.1 | 3907 | UTR 3 | XP_005256978.1 |