Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAGTGAAGAGAGATGCGAGCTG[A/G]CTAGGACAGGGAAGAGAGATGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609949 MIM: 615475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5AR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5AR2 - complement component 5a receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DHX34 - DEAH-box helicase 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014681.5 | Intron | NP_055496.2 | ||||
XM_005259443.3 | Intron | XP_005259500.1 | ||||
XM_011527550.2 | Intron | XP_011525852.1 | ||||
XM_011527551.2 | Intron | XP_011525853.1 | ||||
XM_017027524.1 | Intron | XP_016883013.1 |