Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGACCCAAAGGAAAATAAAAC[C/T]AAAGGCAGAGATATATTTTGGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608792 MIM: 188070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIPC3 - GIPC PDZ domain containing family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXA2R - thromboxane A2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001060.5 | 1661 | UTR 3 | NP_001051.1 | |||
NM_201636.2 | 1661 | UTR 3 | NP_963998.2 | |||
XM_011528214.2 | 1661 | Intron | XP_011526516.1 |