Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCCTGGGCGCGCACTCCCTGT[C/T]GCAGCCGGAGCCCTCCAAGCGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 134350 MIM: 130130 MIM: 604062 | ||||||||||||||||||||
Literature Links: |
CFD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFD - complement factor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317335.1 | 288 | Missense Mutation | TCG,TTG | S,L 93 | NP_001304264.1 | |
NM_001928.3 | 288 | Missense Mutation | TCG,TTG | S,L 86 | NP_001919.2 |
ELANE - elastase, neutrophil expressed | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED16 - mediator complex subunit 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |