Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACGCACAGGGAGAAACGCGTCTC[C/T]CCAAACGGCGGCTGGTGCAGGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A42 - solute carrier family 25 member 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM161A - transmembrane protein 161A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256766.1 | 866 | Silent Mutation | GGA,GGG | G,G 196 | NP_001243695.1 | |
NM_017814.2 | 866 | Silent Mutation | GGA,GGG | G,G 299 | NP_060284.1 | |
XM_011528117.2 | 866 | Silent Mutation | GGA,GGG | G,G 97 | XP_011526419.1 | |
XM_017026916.1 | 866 | Silent Mutation | GGA,GGG | G,G 274 | XP_016882405.1 |