Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACGCCAGTGCATGACCTGACCCT[A/G]ACCTTCACAGCAGCCATTCTCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601052 MIM: 606849 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIN1 - peptidylprolyl cis/trans isomerase, NIMA-interacting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006221.3 | Intron | NP_006212.1 | ||||
XM_011528068.2 | Intron | XP_011526370.1 |
UBL5 - ubiquitin like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |