Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTATTAAATTTGTTTACCTTCT[A/T]AAAAAAACGATTACAAAAAAGAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614610 MIM: 609394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KANK2 - KN motif and ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136191.2 | 5165 | UTR 3 | NP_001129663.1 | |||
NM_015493.6 | 5165 | UTR 3 | NP_056308.3 |
SPC24 - SPC24, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |