Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTTTCCACATTCCTCACAATC[A/G]TAAGGTTTCCCCCCAGTGTGAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194557 | ||||||||||||||||||||
Literature Links: |
ZNF20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF20 - zinc finger protein 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF625 - zinc finger protein 625 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145233.3 | 662 | Silent Mutation | TAC,TAT | Y,Y 163 | NP_660276.2 |
ZNF625-ZNF20 - ZNF625-ZNF20 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |