Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTCGGCTTGAGGATCACTACC[A/G]GCCTCCGCTGTGTCCGTGGGGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP8 - A-kinase anchoring protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005858.3 | 2008 | Silent Mutation | GCC,GCT | A,A 615 | NP_005849.1 | |
XM_011527624.2 | 2008 | Silent Mutation | GCC,GCT | A,A 515 | XP_011525926.1 | |
XM_017026141.1 | 2008 | Silent Mutation | GCC,GCT | A,A 541 | XP_016881630.1 |