Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGAGTTCCCAGGAGCTGGGTGC[G/T]GCGCTAGCGCAGCTGGTGGCCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604951 MIM: 600691 | ||||||||||||||||||||
Literature Links: |
PGLS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGLS - 6-phosphogluconolactonase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012088.2 | 110 | Silent Mutation | GCG,GCT | A,A 20 | NP_036220.1 | |
XM_011527897.2 | 110 | Intron | XP_011526199.1 |
SLC27A1 - solute carrier family 27 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |