Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTGACAACAGTGTACAGTTTT[C/T]CCAGACCCATATGTAGGTTCCATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSFL1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSFL1C - NSFL1 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206736.1 | 4070 | UTR 3 | NP_001193665.1 | |||
NM_016143.4 | 4070 | UTR 3 | NP_057227.2 | |||
NM_018839.4 | 4070 | UTR 3 | NP_061327.2 | |||
XM_006723592.2 | 4070 | UTR 3 | XP_006723655.1 | |||
XM_006723593.2 | 4070 | UTR 3 | XP_006723656.1 | |||
XM_006723594.2 | 4070 | UTR 3 | XP_006723657.1 | |||
XM_011529300.1 | 4070 | UTR 3 | XP_011527602.1 | |||
XM_011529301.1 | 4070 | UTR 3 | XP_011527603.1 | |||
XM_011529302.2 | 4070 | UTR 3 | XP_011527604.1 | |||
XM_011529304.1 | 4070 | UTR 3 | XP_011527606.1 | |||
XM_011529305.2 | 4070 | Intron | XP_011527607.1 | |||
XM_017027956.1 | 4070 | UTR 3 | XP_016883445.1 | |||
XM_017027957.1 | 4070 | UTR 3 | XP_016883446.1 | |||
XM_017027958.1 | 4070 | UTR 3 | XP_016883447.1 |