Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGATGCCTGAATGACTACATGAA[C/T]AAATGAACGGATGAGTCCTTTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610608 MIM: 609580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GINS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GINS1 - GINS complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NINL - ninein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318226.1 | Intron | NP_001305155.1 | ||||
NM_025176.5 | Intron | NP_079452.3 | ||||
XM_011529186.2 | Intron | XP_011527488.1 | ||||
XM_011529187.2 | Intron | XP_011527489.1 | ||||
XM_011529188.1 | Intron | XP_011527490.1 | ||||
XM_011529189.1 | Intron | XP_011527491.1 | ||||
XM_011529190.1 | Intron | XP_011527492.1 | ||||
XM_011529191.1 | Intron | XP_011527493.1 | ||||
XM_011529192.1 | Intron | XP_011527494.1 | ||||
XM_011529193.1 | Intron | XP_011527495.1 | ||||
XM_011529194.2 | Intron | XP_011527496.1 | ||||
XM_011529195.2 | Intron | XP_011527497.1 | ||||
XM_011529197.2 | Intron | XP_011527499.1 | ||||
XM_011529198.1 | Intron | XP_011527500.1 | ||||
XM_017027732.1 | Intron | XP_016883221.1 | ||||
XM_017027733.1 | Intron | XP_016883222.1 | ||||
XM_017027734.1 | Intron | XP_016883223.1 | ||||
XM_017027735.1 | Intron | XP_016883224.1 |