Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCTTGACTTAGAGATGCACAC[A/G]CACACTCACTGCACTTGCTGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605925 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C21orf58 - chromosome 21 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNT - pericentrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001315529.1 | Intron | NP_001302458.1 | ||||
NM_006031.5 | Intron | NP_006022.3 | ||||
XM_005261124.4 | Intron | XP_005261181.1 | ||||
XM_011529594.2 | Intron | XP_011527896.1 | ||||
XM_017028362.1 | Intron | XP_016883851.1 | ||||
XM_017028363.1 | Intron | XP_016883852.1 |