Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCCCCAGATCCTTCTAATGGGG[G/T]AACCTCACCTGCCCTTGTCAGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602705 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYN3 - synapsin III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135774.1 | 5354 | Intron | NP_001129246.1 | |||
NM_003490.3 | 5354 | Intron | NP_003481.3 | |||
NM_133633.2 | 5354 | Intron | NP_598344.2 | |||
XM_011530405.2 | 5354 | UTR 3 | XP_011528707.1 | |||
XM_011530406.2 | 5354 | UTR 3 | XP_011528708.1 | |||
XM_011530407.2 | 5354 | UTR 3 | XP_011528709.1 | |||
XM_011530408.1 | 5354 | UTR 3 | XP_011528710.1 | |||
XM_011530410.2 | 5354 | UTR 3 | XP_011528712.1 | |||
XM_017028960.1 | 5354 | UTR 3 | XP_016884449.1 | |||
XM_017028961.1 | 5354 | UTR 3 | XP_016884450.1 | |||
XM_017028962.1 | 5354 | UTR 3 | XP_016884451.1 | |||
XM_017028963.1 | 5354 | UTR 3 | XP_016884452.1 | |||
XM_017028964.1 | 5354 | UTR 3 | XP_016884453.1 | |||
XM_017028965.1 | 5354 | UTR 3 | XP_016884454.1 |