Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTATTCTGCCTTTCTTGTCTCC[A/T]AAGAATAACTGTGCTTGAAGAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300237 | ||||||||||||||||||||
Literature Links: |
TCEAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCEAL1 - transcription elongation factor A like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006639.1 | Intron | NP_001006640.1 | ||||
NM_001006640.1 | Intron | NP_001006641.1 | ||||
NM_004780.2 | Intron | NP_004771.2 |