Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCGAAGCCATGGCGGGACCTGG[G/T]GGCTGGAGGGACAGGGAGGTCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300791 | ||||||||||||||||||||
Literature Links: |
NRK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRK - Nik related kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198465.3 | 318 | Silent Mutation | GGG,GGT | G,G 5 | NP_940867.2 | |
XM_006724632.3 | 318 | Silent Mutation | GGG,GGT | G,G 5 | XP_006724695.1 | |
XM_006724633.3 | 318 | Silent Mutation | GGG,GGT | G,G 5 | XP_006724696.1 | |
XM_006724634.3 | 318 | Silent Mutation | GGG,GGT | G,G 5 | XP_006724697.1 | |
XM_011530887.2 | 318 | Intron | XP_011529189.1 |