Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCTGGAGCAGAAGAGTCTGCA[A/G]CTAGGAAGGGAACTCTTGTGTGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADH6 - alcohol dehydrogenase 6 (class V) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000672.3 | 2225 | Intron | NP_000663.1 | |||
NM_001102470.1 | 2225 | UTR 3 | NP_001095940.1 |
LOC100507053 - uncharacterized LOC100507053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |