Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGAAAATGCCACATGAAAAGC[A/G]CTATACATTATGTGGAACTCCTAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605031 | ||||||||||||||||||||
Literature Links: |
PLK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLK4 - polo like kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190799.1 | 937 | Missense Mutation | CAC,CGC | H,R 136 | NP_001177728.1 | |
NM_001190801.1 | 937 | Missense Mutation | CAC,CGC | H,R 127 | NP_001177730.1 | |
NM_014264.4 | 937 | Missense Mutation | CAC,CGC | H,R 168 | NP_055079.3 | |
XM_005262701.2 | 937 | Missense Mutation | CAC,CGC | H,R 168 | XP_005262758.1 | |
XM_017007662.1 | 937 | Missense Mutation | CAC,CGC | H,R 169 | XP_016863151.1 | |
XM_017007663.1 | 937 | Missense Mutation | CAC,CGC | H,R 169 | XP_016863152.1 |