Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATAACAATTTCACTTGGGAATG[G/T]TTTTTTTTAAAAAATGACAAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAP130 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAP130 - Sin3A associated protein 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145928.1 | 3825 | UTR 3 | NP_001139400.1 | |||
NM_024545.3 | 3825 | UTR 3 | NP_078821.2 | |||
XM_005263767.3 | 3825 | UTR 3 | XP_005263824.1 | |||
XM_005263768.3 | 3825 | UTR 3 | XP_005263825.1 | |||
XM_005263769.3 | 3825 | UTR 3 | XP_005263826.1 | |||
XM_005263770.3 | 3825 | UTR 3 | XP_005263827.1 | |||
XM_006712748.3 | 3825 | UTR 3 | XP_006712811.1 | |||
XM_006712749.3 | 3825 | UTR 3 | XP_006712812.1 | |||
XM_017004903.1 | 3825 | UTR 3 | XP_016860392.1 | |||
XM_017004904.1 | 3825 | UTR 3 | XP_016860393.1 | |||
XM_017004905.1 | 3825 | UTR 3 | XP_016860394.1 |