Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATATTTTACTTTTTATAACTTT[A/G]GAAACCTTAGAAGTTGCCAATTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPC2 - enhancer of polycomb homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015630.3 | Intron | NP_056445.3 | ||||
XM_011510941.2 | Intron | XP_011509243.1 | ||||
XM_011510943.2 | Intron | XP_011509245.1 |