Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCATATCCTGCAGTTTGAAAGT[A/G]CATTTTTCATAATTACTGGATTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605638 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIRC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BIRC6 - baculoviral IAP repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016252.3 | Intron | NP_057336.3 | ||||
XM_005264449.4 | Intron | XP_005264506.2 | ||||
XM_005264450.4 | Intron | XP_005264507.2 | ||||
XM_005264451.4 | Intron | XP_005264508.2 | ||||
XM_005264452.4 | Intron | XP_005264509.2 | ||||
XM_005264453.4 | Intron | XP_005264510.2 | ||||
XM_005264454.4 | Intron | XP_005264511.2 | ||||
XM_005264455.4 | Intron | XP_005264512.2 | ||||
XM_006712054.3 | Intron | XP_006712117.1 | ||||
XM_006712055.3 | Intron | XP_006712118.1 | ||||
XM_006712056.3 | Intron | XP_006712119.1 | ||||
XM_011533003.2 | Intron | XP_011531305.1 | ||||
XM_011533005.2 | Intron | XP_011531307.1 | ||||
XM_017004556.1 | Intron | XP_016860045.1 | ||||
XM_017004557.1 | Intron | XP_016860046.1 | ||||
XM_017004558.1 | Intron | XP_016860047.1 | ||||
XM_017004559.1 | Intron | XP_016860048.1 | ||||
XM_017004560.1 | Intron | XP_016860049.1 |