Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCAGGTGGTGTCGGGCGAGTTC[A/T]GCGAGGACAGCGAGGTGTACAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAREM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAREM2 - GRB2 associated regulator of MAPK1 subtype 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168241.1 | 543 | Missense Mutation | AGC,TGC | S,C 135 | NP_001161713.1 | |
NM_001191033.1 | 543 | Missense Mutation | AGC,TGC | S,C 58 | NP_001177962.1 | |
XM_006711951.3 | 543 | Missense Mutation | AGC,TGC | S,C 135 | XP_006712014.1 | |
XM_011532564.2 | 543 | Missense Mutation | AGC,TGC | S,C 133 | XP_011530866.1 | |
XM_011532565.2 | 543 | Missense Mutation | AGC,TGC | S,C 133 | XP_011530867.1 | |
XM_011532566.2 | 543 | Missense Mutation | AGC,TGC | S,C 16 | XP_011530868.1 | |
XM_011532567.2 | 543 | Missense Mutation | AGC,TGC | S,C 135 | XP_011530869.1 |
HADHA - hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |