Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGTACTACAGTGAGTTTTGAC[A/C]AACCAACCATTTCGTGTGTGTGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108733 MIM: 600152 | ||||||||||||||||||||
Literature Links: |
ATP2B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP2B2 - ATPase plasma membrane Ca2+ transporting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001331.2 | 7051 | UTR 3 | NP_001001331.1 | |||
NM_001683.3 | 7051 | UTR 3 | NP_001674.2 | |||
XM_005265179.4 | 7051 | UTR 3 | XP_005265236.1 | |||
XM_006713175.3 | 7051 | UTR 3 | XP_006713238.1 | |||
XM_011533752.2 | 7051 | UTR 3 | XP_011532054.1 | |||
XM_017006481.1 | 7051 | UTR 3 | XP_016861970.1 | |||
XM_017006482.1 | 7051 | UTR 3 | XP_016861971.1 | |||
XM_017006483.1 | 7051 | UTR 3 | XP_016861972.1 | |||
XM_017006484.1 | 7051 | UTR 3 | XP_016861973.1 | |||
XM_017006485.1 | 7051 | UTR 3 | XP_016861974.1 | |||
XM_017006486.1 | 7051 | UTR 3 | XP_016861975.1 | |||
XM_017006487.1 | 7051 | UTR 3 | XP_016861976.1 | |||
XM_017006488.1 | 7051 | Intron | XP_016861977.1 | |||
XM_017006489.1 | 7051 | UTR 3 | XP_016861978.1 | |||
XM_017006490.1 | 7051 | Intron | XP_016861979.1 | |||
XM_017006491.1 | 7051 | UTR 3 | XP_016861980.1 | |||
XM_017006492.1 | 7051 | UTR 3 | XP_016861981.1 |
MIR378B - microRNA 378b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC13 - SEC13 homolog, nuclear pore and COPII coat complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |