Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAGAAAATCAACACTTTTGAGT[A/G]GAAAATCCGGTAGAGAAAAGACCTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF385D PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF385D - zinc finger protein 385D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024697.2 | Intron | NP_078973.1 | ||||
XM_011534122.1 | Intron | XP_011532424.1 | ||||
XM_011534123.2 | Intron | XP_011532425.1 | ||||
XM_011534124.2 | Intron | XP_011532426.1 | ||||
XM_017007191.1 | Intron | XP_016862680.1 | ||||
XM_017007192.1 | Intron | XP_016862681.1 | ||||
XM_017007193.1 | Intron | XP_016862682.1 | ||||
XM_017007194.1 | Intron | XP_016862683.1 | ||||
XM_017007195.1 | Intron | XP_016862684.1 | ||||
XM_017007196.1 | Intron | XP_016862685.1 | ||||
XM_017007197.1 | Intron | XP_016862686.1 | ||||
XM_017007198.1 | Intron | XP_016862687.1 | ||||
XM_017007199.1 | Intron | XP_016862688.1 | ||||
XM_017007200.1 | Intron | XP_016862689.1 | ||||
XM_017007201.1 | Intron | XP_016862690.1 | ||||
XM_017007202.1 | Intron | XP_016862691.1 | ||||
XM_017007203.1 | Intron | XP_016862692.1 |