Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGGACTAAGCAAATTTTCCAC[A/G]TTGCTATCCTTCGCTATCAGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308173.1 | 2269 | UTR 3 | NP_001295102.1 | |||
NM_024565.6 | 2269 | UTR 3 | NP_078841.3 | |||
XM_006714917.3 | 2269 | UTR 3 | XP_006714980.1 | |||
XM_011534646.2 | 2269 | UTR 3 | XP_011532948.1 | |||
XM_011534647.2 | 2269 | UTR 3 | XP_011532949.1 | |||
XM_017009846.1 | 2269 | UTR 3 | XP_016865335.1 | |||
XM_017009847.1 | 2269 | UTR 3 | XP_016865336.1 | |||
XM_017009848.1 | 2269 | UTR 3 | XP_016865337.1 | |||
XM_017009849.1 | 2269 | UTR 3 | XP_016865338.1 |