Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGATGTGAGGAGGTAATAGAGGT[A/G]AGAGAGCATATATTACAGTATATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP18 - Rho GTPase activating protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033515.2 | 2328 | UTR 3 | NP_277050.2 | |||
XM_005267212.2 | 2328 | UTR 3 | XP_005267269.2 | |||
XM_005267213.1 | 2328 | UTR 3 | XP_005267270.1 | |||
XM_017011494.1 | 2328 | UTR 3 | XP_016866983.1 |