Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCTTCTTCCACACATGCAAGG[G/T]GAAAGGGTGTGGATTTTAAGACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176851 | ||||||||||||||||||||
Literature Links: |
PCMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCMT1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252049.1 | 930 | Intron | NP_001238978.1 | |||
NM_001252050.1 | 930 | Intron | NP_001238979.1 | |||
NM_001252051.1 | 930 | Intron | NP_001238980.1 | |||
NM_001252052.1 | 930 | Intron | NP_001238981.1 | |||
NM_001252053.1 | 930 | UTR 3 | NP_001238982.1 | |||
NM_005389.2 | 930 | Intron | NP_005380.2 | |||
XM_011535868.2 | 930 | Intron | XP_011534170.1 |