Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGATATGCATGCGTGCTGTCT[C/T]ATTGTTGCAAAAATGAGTCTTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601554 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNLT1 - dynein light chain Tctex-type 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL3 - synaptotagmin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009991.3 | Intron | NP_001009991.2 | ||||
NM_001242384.1 | Intron | NP_001229313.1 | ||||
NM_001242394.1 | Intron | NP_001229323.1 | ||||
NM_001242395.1 | Intron | NP_001229324.1 | ||||
NM_001318745.1 | Intron | NP_001305674.1 | ||||
XM_005267215.4 | Intron | XP_005267272.1 | ||||
XM_005267218.4 | Intron | XP_005267275.1 | ||||
XM_005267222.4 | Intron | XP_005267279.1 | ||||
XM_006715605.3 | Intron | XP_006715668.1 | ||||
XM_006715606.3 | Intron | XP_006715669.1 | ||||
XM_006715611.3 | Intron | XP_006715674.1 | ||||
XM_011536254.2 | Intron | XP_011534556.1 | ||||
XM_011536255.2 | Intron | XP_011534557.1 | ||||
XM_017011495.1 | Intron | XP_016866984.1 | ||||
XM_017011496.1 | Intron | XP_016866985.1 | ||||
XM_017011497.1 | Intron | XP_016866986.1 | ||||
XM_017011498.1 | Intron | XP_016866987.1 | ||||
XM_017011499.1 | Intron | XP_016866988.1 |