Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGAAAAAAGGCGGGAGATGTAG[C/T]GATGCTGAGCAAAGAGCAGATATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605824 | ||||||||||||||||||||
Literature Links: |
BVES-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BVES-AS1 - BVES antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POPDC3 - popeye domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022361.4 | 1979 | Missense Mutation | CAC,CGC | H,R 225 | NP_071756.2 | |
XM_011536067.2 | 1979 | Missense Mutation | CAC,CGC | H,R 225 | XP_011534369.1 | |
XM_017011194.1 | 1979 | Missense Mutation | CAC,CGC | H,R 225 | XP_016866683.1 | |
XM_017011195.1 | 1979 | Missense Mutation | CAC,CGC | H,R 225 | XP_016866684.1 |