Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATGAGAGACCAAACGAGCTTAG[C/G]TAGCGTAATGCTAAAAAATTGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
L2HGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
L2HGDH - L-2-hydroxyglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024884.2 | 1904 | UTR 3 | NP_079160.1 | |||
XM_005268075.4 | 1904 | UTR 3 | XP_005268132.1 | |||
XM_011537166.2 | 1904 | Intron | XP_011535468.1 | |||
XM_011537167.2 | 1904 | Intron | XP_011535469.1 | |||
XM_011537168.2 | 1904 | Intron | XP_011535470.1 | |||
XM_017021655.1 | 1904 | Intron | XP_016877144.1 | |||
XM_017021656.1 | 1904 | Intron | XP_016877145.1 | |||
XM_017021657.1 | 1904 | UTR 3 | XP_016877146.1 | |||
XM_017021658.1 | 1904 | Intron | XP_016877147.1 |