Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATATTTGAGGGAAGAAAACAGC[A/G]AAGTATGTATATTCTGGGTTTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TC2N PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128595.2 | 2379 | UTR 3 | NP_001122067.1 | |||
NM_001128596.2 | 2379 | UTR 3 | NP_001122068.1 | |||
NM_001289134.1 | 2379 | UTR 3 | NP_001276063.1 | |||
NM_152332.5 | 2379 | UTR 3 | NP_689545.1 |