Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTCAGCACAACCCCAGCACTG[C/T]TGTCAGGTAACGCTGGCAAAGGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604505 | ||||||||||||||||||||
Literature Links: |
TRIP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIP11 - thyroid hormone receptor interactor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321851.1 | 6295 | Missense Mutation | AAC,AGC | N,S 1966 | NP_001308780.1 | |
NM_004239.4 | 6295 | Missense Mutation | AAC,AGC | N,S 1967 | NP_004230.2 | |
XM_017021787.1 | 6295 | Missense Mutation | AAC,AGC | N,S 1732 | XP_016877276.1 | |
XM_017021788.1 | 6295 | Missense Mutation | AAC,AGC | N,S 1525 | XP_016877277.1 |