Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAGCTCAGGCTCTTGGGACAT[C/T]GTGGAGGTACTGGGCACCGCTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612374 | ||||||||||||||||||||
Literature Links: |
TMEM173 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM173 - transmembrane protein 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301738.1 | 1145 | UTR 3 | NP_001288667.1 | |||
NM_198282.3 | 1145 | Silent Mutation | ACC,ACT | T,T 356 | NP_938023.1 | |
XM_005268445.3 | 1145 | UTR 3 | XP_005268502.1 | |||
XM_011537639.2 | 1145 | Intron | XP_011535941.1 | |||
XM_011537640.2 | 1145 | Silent Mutation | ACC,ACT | T,T 237 | XP_011535942.1 |