Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGAGTAGGGTTAAAAGAGGC[C/T]TTTGTAGTTCTTCATTGAAGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNOT2 - CCR4-NOT transcription complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199302.1 | Intron | NP_001186231.1 | ||||
NM_001199303.1 | Intron | NP_001186232.1 | ||||
NM_014515.5 | Intron | NP_055330.1 | ||||
XM_006719429.3 | Intron | XP_006719492.1 | ||||
XM_006719430.3 | Intron | XP_006719493.1 | ||||
XM_006719432.3 | Intron | XP_006719495.1 | ||||
XM_006719433.3 | Intron | XP_006719496.1 | ||||
XM_011538399.1 | Intron | XP_011536701.1 | ||||
XM_011538400.2 | Intron | XP_011536702.1 | ||||
XM_017019348.1 | Intron | XP_016874837.1 | ||||
XM_017019349.1 | Intron | XP_016874838.1 | ||||
XM_017019350.1 | Intron | XP_016874839.1 | ||||
XM_017019351.1 | Intron | XP_016874840.1 | ||||
XM_017019352.1 | Intron | XP_016874841.1 | ||||
XM_017019353.1 | Intron | XP_016874842.1 | ||||
XM_017019354.1 | Intron | XP_016874843.1 |
LINC01481 - long intergenic non-protein coding RNA 1481 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |