Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTTAATACATATTCTTTAGGAA[C/T]AGTTGCTAACTAACTCATAACTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616747 MIM: 604759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHPT1 - choline phosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020244.2 | 1029 | Intron | NP_064629.2 | |||
XM_011538574.1 | 1029 | Intron | XP_011536876.1 | |||
XM_011538575.1 | 1029 | Intron | XP_011536877.1 |
SYCP3 - synaptonemal complex protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177948.1 | 1029 | UTR 3 | NP_001171419.1 | |||
NM_001177949.1 | 1029 | UTR 3 | NP_001171420.1 | |||
NM_153694.4 | 1029 | UTR 3 | NP_710161.1 | |||
XM_005268922.4 | 1029 | UTR 3 | XP_005268979.3 | |||
XM_005268924.1 | 1029 | UTR 3 | XP_005268981.1 | |||
XM_005268925.1 | 1029 | UTR 3 | XP_005268982.1 | |||
XM_005268926.3 | 1029 | UTR 3 | XP_005268983.1 | |||
XM_005268927.2 | 1029 | UTR 3 | XP_005268984.1 | |||
XM_011538421.2 | 1029 | UTR 3 | XP_011536723.2 | |||
XM_017019368.1 | 1029 | UTR 3 | XP_016874857.1 |