Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTAGTGACACATTCAAGTGAGGC[A/T]GTAAATCTAGGTAAGTGACACTAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 190010 | |||||||||||||||||||||||
Literature Links: |
TFRC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TFRC - transferrin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128148.2 | 4108 | UTR 3 | NP_001121620.1 | |||
NM_001313965.1 | 4108 | UTR 3 | NP_001300894.1 | |||
NM_001313966.1 | 4108 | UTR 3 | NP_001300895.1 | |||
NM_003234.3 | 4108 | UTR 3 | NP_003225.2 |