Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGACGGAGTCCGTGGTGTCCCG[A/T]TGTAACTGACCCCTGCTCCAAACGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606871 MIM: 609276 | ||||||||||||||||||||
Literature Links: |
JAM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JAM3 - junctional adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPD3 - non-SMC condensin II complex subunit D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015261.2 | 4901 | Missense Mutation | AAC,ATC | N,I 1432 | NP_056076.1 | |
XM_011542719.2 | 4901 | Missense Mutation | AAC,ATC | N,I 1294 | XP_011541021.1 |