Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCTCATCGGCATCACGGGCCA[C/T]GGCATCCCCTGGATCGGAGGCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM78A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM78A - family with sequence similarity 78 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLPP7 - phospholipid phosphatase 7 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032728.3 | 679 | Silent Mutation | CAC,CAT | H,H 121 | NP_116117.3 | |
XM_005272230.3 | 679 | Silent Mutation | CAC,CAT | H,H 121 | XP_005272287.1 | |
XM_005272231.1 | 679 | Silent Mutation | CAC,CAT | H,H 121 | XP_005272288.1 |