Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGATTTAAATATTGCTACTCTT[C/T]AGGGTCATTGCTCTACCTACAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164831 MIM: 616700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMI1 - BMI1 proto-oncogene, polycomb ring finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005180.8 | Intron | NP_005171.4 |
COMMD3 - COMM domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COMMD3-BMI1 - COMMD3-BMI1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204062.1 | Intron | NP_001190991.1 |