Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGCTACCTGCTCTTCAATGACA[C/T]TGATCTGATCCAGATAAGGCTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609768 MIM: 604532 | ||||||||||||||||||||
Literature Links: |
BLOC1S2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BLOC1S2 - biogenesis of lysosomal organelles complex 1 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001342.2 | 478 | Missense Mutation | AAT,AGT | N,S 66 | NP_001001342.1 | |
NM_001282436.1 | 478 | Missense Mutation | AAT,AGT | N,S 62 | NP_001269365.1 | |
NM_001282437.1 | 478 | Missense Mutation | AAT,AGT | N,S 66 | NP_001269366.1 | |
NM_001282438.1 | 478 | Missense Mutation | AAT,AGT | N,S 66 | NP_001269367.1 | |
NM_001282439.1 | 478 | Missense Mutation | AAT,AGT | N,S 109 | NP_001269368.1 | |
NM_173809.4 | 478 | Missense Mutation | AAT,AGT | N,S 109 | NP_776170.2 | |
XM_017016102.1 | 478 | Missense Mutation | AAT,AGT | N,S 66 | XP_016871591.1 |
PKD2L1 - polycystin 2 like 1, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |