Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATGGAGGATCTGCTGTCTCCGT[A/G]TGCGTAGCCTTGGATTCCAGTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607318 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOXL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOXL4 - lysyl oxidase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032211.6 | Intron | NP_115587.6 | ||||
XM_005270216.1 | Intron | XP_005270273.1 | ||||
XM_006718020.1 | Intron | XP_006718083.1 |
R3HCC1L - R3H domain and coiled-coil containing 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |