Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGCAGAAGCCCAGACTTACCTG[C/G]ACAAGGTGGAGAACATTTGCAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147690 | ||||||||||||||||||||
Literature Links: |
IFIT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFIT1 - interferon induced protein with tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270927.1 | 631 | Missense Mutation | CAC,GAC | H,D 119 | NP_001257856.1 | |
NM_001270928.1 | 631 | Missense Mutation | CAC,GAC | H,D 88 | NP_001257857.1 | |
NM_001270929.1 | 631 | Missense Mutation | CAC,GAC | H,D 88 | NP_001257858.1 | |
NM_001270930.1 | 631 | Missense Mutation | CAC,GAC | H,D 88 | NP_001257859.1 | |
NM_001548.4 | 631 | Missense Mutation | CAC,GAC | H,D 119 | NP_001539.3 |