Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCTTTTTTCCAGTCTGATGGC[C/T]CATACCTCCTTGGCCCTCTTTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EBF3 - early B-cell factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005463.2 | 1901 | UTR 3 | NP_001005463.1 | |||
XM_005252667.3 | 1901 | UTR 3 | XP_005252724.1 | |||
XM_005252668.3 | 1901 | UTR 3 | XP_005252725.1 | |||
XM_005252669.3 | 1901 | UTR 3 | XP_005252726.1 | |||
XM_006717739.3 | 1901 | UTR 3 | XP_006717802.1 | |||
XM_006717740.3 | 1901 | UTR 3 | XP_006717803.1 | |||
XM_006717741.3 | 1901 | Intron | XP_006717804.1 | |||
XM_006717742.3 | 1901 | UTR 3 | XP_006717805.1 | |||
XM_006717743.3 | 1901 | UTR 3 | XP_006717806.1 | |||
XM_006717744.3 | 1901 | UTR 3 | XP_006717807.1 | |||
XM_011539574.2 | 1901 | UTR 3 | XP_011537876.1 | |||
XM_011539575.2 | 1901 | UTR 3 | XP_011537877.1 | |||
XM_017016027.1 | 1901 | UTR 3 | XP_016871516.1 |
MIR4297 - microRNA 4297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |