Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGTTTTTCAACAGCTTAATGA[A/T]TGAAGTGGTTCATACTTCTCCAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608909 | ||||||||||||||||||||
Literature Links: |
ARL5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL5B - ADP ribosylation factor like GTPase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178815.3 | 419 | Missense Mutation | AAT,ATT | N,I 39 | NP_848930.1 | |
XM_005252400.1 | 419 | Missense Mutation | AAT,ATT | N,I 39 | XP_005252457.1 | |
XM_005252401.4 | 419 | Missense Mutation | AAT,ATT | N,I 2 | XP_005252458.1 |
NSUN6 - NOP2/Sun RNA methyltransferase family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |